Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200R1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580280 Supplier Invitrogen™ Supplier No. PA580280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Cynomolgus CD200RLa.

May be a receptor for the CD200/OX2 cell surface glycoprotein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R1L
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD200R1L
Gene Accession No. 0
Gene Alias CD200 cell surface glycoprotein receptor 2; CD200 cell surface glycoprotein receptor-like 2; CD200 cell surface glycoprotein receptor-like a; CD200 receptor 1 like; CD200 receptor 1-like; CD200 receptor 2; CD200 receptor-like 2; CD200R1L; CD200R2; CD200RLa; Cell surface glycoprotein CD200 receptor 1-like; cell surface glycoprotein CD200 receptor 2; Cell surface glycoprotein OX2 receptor 2; huCD200R2
Gene Symbols CD200R1L
Host Species Rabbit
Immunogen Recombinant Cynomolgus CD200RLa protein.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102140606
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.