Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD200R1L Recombinant Rabbit Monoclonal Antibody (015), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543068
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543068 100 μL
PIMA543465 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543068 Supplier Invitrogen™ Supplier No. MA543068
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Cynomolgus CD200RLa

May be a receptor for the CD200/OX2 cell surface glycoprotein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD200R1L
Applications ELISA
Classification Recombinant Monoclonal
Clone 015
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD200R1L
Gene Accession No. 0
Gene Alias CD200 cell surface glycoprotein receptor 2; CD200 cell surface glycoprotein receptor-like 2; CD200 cell surface glycoprotein receptor-like a; CD200 receptor 1 like; CD200 receptor 1-like; CD200 receptor 2; CD200 receptor-like 2; CD200R1L; CD200R2; CD200RLa; Cell surface glycoprotein CD200 receptor 1-like; cell surface glycoprotein CD200 receptor 2; Cell surface glycoprotein OX2 receptor 2; huCD200R2
Gene Symbols CD200R1L
Host Species Rabbit
Immunogen Recombinant Cynomolgus CD200RLa protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102140606
Target Species Cynomolgus Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.