Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R4 Antibody (4), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290661AF700
Description
CD200R4 Monoclonal specifically detects CD200R4 in Mouse samples. It is validated for ELISA.Specifications
CD200R4 | |
Monoclonal | |
Alexa Fluor 700 | |
F630107N04Rik, MCD200, MCD200RLa | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R4 (NP_997127.1; Met 1-Thr 241). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
4 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
239849 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction