Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306093AF350
Description
CD200R4 Polyclonal specifically detects CD200R4 in Mouse samples. It is validated for ELISA.Specifications
CD200R4 | |
Polyclonal | |
ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
239849 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Produced in rabbits immunized with purified, recombinant Mouse CD200R4 (NP_997127.1; Met 1-Thr 241). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction