Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R4 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306093APC
Description
CD200R4 Polyclonal antibody specifically detects CD200R4 in Mouse samples. It is validated for ELISASpecifications
CD200R4 | |
Polyclonal | |
ELISA | |
F630107N04Rik, MCD200, MCD200RLa | |
Produced in rabbits immunized with purified, recombinant Mouse CD200R4 (Accession#: NP_997127.1; Met 1-Thr 241) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
239849 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction