Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cd200r4 Recombinant Rabbit Monoclonal Antibody (4)
GREENER_CHOICE

Catalog No. PIMA530238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530238 Supplier Invitrogen™ Supplier No. MA530238
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse CD200R4/CD200 Receptor 4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Cd200r4
Applications ELISA
Classification Recombinant Monoclonal
Clone 4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Cd200r4
Gene Accession No. Q6XJV4
Gene Alias CD200 cell surface glycoprotein receptor; CD200 cell surface glycoprotein receptor-like 4; CD200 cell surface glycoprotein receptor-like a; CD200 receptor 4; CD200 receptor-like 4; Cd200r4; CD200RLa; cell surface glycoprotein CD200 receptor 4; Cell surface glycoprotein OX2 receptor 4; F630107N04Rik; MCD200RLa
Gene Symbols Cd200r4
Host Species Rabbit
Immunogen Recombinant Mouse CD200R4 protein (Met1-Thr241).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239849
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.