Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD201 (EPCR) Monoclonal Antibody (eBiomRCR-16 (mRCR-16)), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. 5014496
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-144-96 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-144-96 Supplier Invitrogen™ Supplier No. 16201181
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The eBioRCR-16 monoclonal antibody reacts with mouse Endothelial Protein C Receptor (EPCR, CD201), a 25 kDa Type 1 transmembrane protein expressed on endothelial cells. EPCR exhibits sequence and structural homology with the MHC class I/CD1 family of proteins. EPCR is a ligand for Protein C and plays an important role in augmenting Protein C activation by the thrombin-thrombomodulin complex and in regulating blood coagulation and inflammation. Deletion of EPCR results in embryonic lethality, at least partially due to placental thrombosis. Recently, it was demonstrated that EPCR expression identified cells in the bone marrow that are capable of hematopoietic reconstitution activity comparable to hematopoietic stem cells isolated with conventional methods. The eBio1560 monoclonal antibody can be used for the detection of these hematopoietic stem cells, however the eBiomRCR-16 monoclonal antibody should only be used for the detection of CD201 on endothelial cells. Applications Reported: This eBiomRCR-16 (mRCR-16) antibody has been reported for use in flow cytometric analysis and in functional assays. Applications Tested: This eBiomRCR-16 (mRCR-16) antibody has been tested by flow cytometric analyis of the bEnd.3 cell line. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^...

The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD201 (EPCR)
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone eBiomRCR-16 (mRCR-16)
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Procr
Gene Accession No. Q64695
Gene Alias Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial
Gene Symbols Procr
Host Species Rat
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19124
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.