Learn More
CD201 Rat anti-Human, Brilliant Violet 605, Clone: RCR-252, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 743553
Description
The RCR-252 monoclonal antibody specifically binds to the endothelial cell protein C receptor (EPCR), which is also known as CD201. CD201 is a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. CD201 binds activated protein C, a serine protease that is involved in regulating blood coagulation. EPCR is reportedly expressed on several malignant cell lines. Malignant cell lines that express EPCR can show significant levels of protein C activation and this activation can be inhibited by anti-EPCR antiboy.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
| CD201 | |
| RCR-252 | |
| Brilliant Violet 605 | |
| EPCR, Activated protein C receptor, APC receptor, PROCR, centrocyclin | |
| EPCR-positive RE-1 Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.