Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD202b (TIE2) Monoclonal Antibody (TEK4), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7069505
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-134-12 100 μg
50-134-13 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-134-12 Supplier Invitrogen™ Supplier No. 14598782
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells. Applications Reported: This TEK4 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: The TEK4 antibody has been tested by blocking of staining with fluorochrome conjugated TEK4 on mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD202b (TIE2)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TEK4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TEK
Gene Accession No. Q02858
Gene Alias AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1
Gene Symbols TEK
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21687
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.