Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD203a Monoclonal Antibody (PM18-7)
GREENER_CHOICE

Catalog No. PIMA528313
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528313 Supplier Invitrogen™ Supplier No. MA528313
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes porcine CD203a, originally clustered as SWC9 at the Second International Swine CD Workshop and later identified as the porcine homologue of human ecto-nucleotidepyrophosphatase/phosphodiesterase 1 or ENPP1. SWC1a, expressed at very much higher levels on monocytes than mature macrophages and CD203a (SWC9), expressed exclusively on mature tissue macrophages have been used as markers of monocyte-macrophage differentiation.

CD203a, originally clustered as SWC9 at the Second International Swine CD Workshop was later identified as the porcine homologue of human ecto-nucleotidepyrophosphatase/phosphodiesterase 1 or ENPP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD203a
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone PM18-7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias CD203; SWC9
Host Species Mouse
Immunogen Porcine alveolar macrophages.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.