Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD203c Monoclonal Antibody (NP4D6), PE
GREENER_CHOICE

Catalog No. PIMA110139
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110139 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110139 Supplier Invitrogen™ Supplier No. MA110139
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with an extracellular epitope of CD203c, a transmembrane ectoenzyme expressed on basophils and mast cells, and overexpressed upon their activation.

The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD203c
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone NP4D6
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene ENPP3
Gene Accession No. O14638
Gene Alias AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen
Gene Symbols ENPP3
Host Species Mouse
Immunogen HEK-293 cells transfected with human CD203c.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5169
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.