Learn More
Invitrogen™ CD203c Monoclonal Antibody (NP4D6), PE, eBioscience™
Mouse Monoclonal Antibody
$398.00
Specifications
Antigen | CD203c |
---|---|
Clone | NP4D6 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-203-942
![]() |
Invitrogen™
12203942 |
100 Tests |
Each of 1 for $398.00
|
![]() |
|||||
Description
Description: This NP4D6 monoclonal antibody reacts with human CD203c, which is also known as ectonucleotide pyrophosphatase/phosphodiesterase 3 (E-NPP3). ENPP family members catalyze the hydrolysis of pyrophosphate and phosphodiester bonds to nucleoside 5-monophosphates. A type II transmembrane protein, CD203c is expressed on basophils and mast cells. CD203c expression is upregulated on basophils activated by allergen or IgE crosslinking. Therefore, CD203c is used as an indicator of basophil activation in various IgE-mediated allergic responses. Applications Reported: This NP4D6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NP4D6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008].Specifications
CD203c | |
5 μL/Test | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
O14638 | |
5169 | |
Primary | |
Affinity Chromatography |
NP4D6 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
ENPP3 | |
AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen | |
ENPP3 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.