Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD203c Mouse anti-Human, Brilliant Violet 650, Clone: NP4D6, BD Optibuild™

Catalog No. BDB744243 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744243 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744243 Supplier BD Biosciences Supplier No. 744243
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The NP4D6 monoclonal antibody specifically binds to CD203c. CD203c is a type II transmembrane glycoprotein that is a member of the E-NNP family of ectoenzymes. CD203c is also known as ectonucleotide pyrophosphatase/phosphodiesterase 3 (E-NPP3, ENPP3) due to its capacity to hydrolyze phosphodiester and phosphosulfate bonds in a variety of molecules including deoxynucleotides, nucleoside phosphates, and nicotinamide adenine dinucleotide. CD203c is otherwise known as Phosphodiesterase-1 (PD-1, PD-I beta), B10, or gp130RB13-6. CD203c is expressed by basophils and mast cells. Basophils increase CD203c expression following activation with allergens or antibodies that crosslink cytophilic IgE. Thus, CD203c serves as a useful flow cytometric marker for basophil activation and the detection and analysis of type 1 allergic responses. IgE-receptor cross-linking results in CD203c upregulation and overexpression on neoplastic mast cells in cases of systemic mastocytosis.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD203c
Classification Monoclonal
Clone NP4D6
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ENPP3, E-NPP 3, NPP3, PDNP3, PD-Ibeta, PD-1β, B10, NPPase, gp130RB13-6
Host Species Mouse
Immunogen Human CD203c Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.