Learn More
Invitrogen™ CD204 Monoclonal Antibody (2F8), PE

Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD204 antibody, clone 2F8 recognizes the murine scavenger receptor class A (SRA), type I and II, also known as CD204.
Specifications
Specifications
| Antigen | CD204 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2F8 |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 1% BSA, 5% sucrose and 0.09% sodium azide |
| Gene | MSR1 |
| Gene Accession No. | P30204 |
| Gene Alias | bovine scavenger receptor type II; CD204; Macrophage acetylated LDL receptor I and II; macrophage scavenger receptor 1; macrophage scavenger receptor type III; macrophage scavenger receptor types I and II; MRS-A; MSR; Msr1; MSR-A; phSR1; phSR2; RGD1564316; SCARA1; Scavenger receptor class A member 1; scavenger receptor class A, member 1; scavenger receptor type A; scavenger receptor type I; Scvr; SRA; SR-A; SR-AI; SR-AI/II; SR-AII |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.