Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD205 (DEC-205) Rat anti-Mouse, Brilliant Violet 421, Clone: V18-949, BD

Catalog No. BDB566375 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566375 100 μg
BDB566376 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566375 Supplier BD Biosciences Supplier No. 566375
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The V18-949 monoclonal antibody specifically recognizes CD205 which is also known as Dendritic and thymic Epithelial Cells, 205 kDa (DEC-205). CD205 (DEC-205) is a type 1 transmembrane glycoprotein that is encoded by Ly75 (lymphocyte antigen 75). CD205 (DEC-205) is highly expressed by both CD8+ and CD103+ dendritic cell subsets and Langerhans cells. It is variably expressed by some other dendritic cell types, Gr1+ (Ly-6G and Ly-6C) bone marrow cells, B cells, macrophages, granulocytes, and mucosal and thymic cortical epithelial cells. CD205 (DEC-205) belongs to the C-type lectin receptor family and functions as a pattern recognition receptor and an endocytic receptor that participates in the cellular processing and presentation of foreign or self antigens to initiate immunity or tolerance.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD205 (DEC-205)
Classification Monoclonal
Clone V18-949
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Cd205, DEC-205, DEC205, Ly-75, Ly75, Lymphocyte antigen 75
Host Species Rat
Immunogen Mouse CD205 (DEC-205)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.