Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD205 (DEC-205) Rat anti-Mouse, BV 711, Clone: NLDC-145, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 752385
This item is not returnable.
View return policy
Description
CD205 (DEC-205) Monoclonal antibody specifically detects CD205 (DEC-205) in Mouse samples. It is validated for Flow Cytometry
Specifications
CD205 (DEC-205) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Flow Cytometry | |
NLDC-145 | |
Brilliant Violet 711 | |
CD205; Cd205; DEC-205; DEC205; Ly-75; Ly75 | |
Mouse lymph node stromal cells | |
RUO | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction