Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD205 Monoclonal Antibody (205yekta), Alexa Fluor™ 488, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

$370.00

Specifications

Antigen CD205
Clone 205yekta
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-246-076
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
53205182
100 μg
Each of 1 for $370.00
Only null left
Add to Cart
 
Description

Description

Description: The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells. Applications Reported: This 205yekta antibody has been reported for use in flow cytometric analysis. Applications Tested: This 205yekta antibody has been tested by flwo cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.
Specifications

Specifications

CD205
0.5 mg/mL
Monoclonal
Liquid
RUO
Q60767
IgG2a κ
Affinity chromatography
Antibody
205yekta
Flow Cytometry
Alexa Fluor 488
Rat
Mouse
17076
Primary
4°C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.