Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD205 Monoclonal Antibody (205yekta), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$201.00 - $379.00
Specifications
Antigen | CD205 |
---|---|
Clone | 205yekta |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M051T03Y03A
![]() |
Invitrogen™
M051T03Y03A |
100 μg |
Each for $379.00
|
|
|||||
M051T02Y03A
![]() |
Invitrogen™
M051T02Y03A |
25 μg |
Each for $201.00
|
|
|||||
Description
CD205 Monoclonal antibody specifically detects CD205 in Mouse samples. It is validated for Flow CytometrySpecifications
CD205 | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Q60767 | |
17076 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
205yekta | |
Flow Cytometry | |
NovaFluor Yellow 610 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4 | |
IgG2a κ | |
Affinity chromatography | |
LY75 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title