Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD205 Monoclonal Antibody (205yekta), Super Bright™ 600, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$478.00
Specifications
| Antigen | CD205 |
|---|---|
| Clone | 205yekta |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
63-205-182
![]()
|
Invitrogen™
63205182 |
100 μg |
Each of 1 for $478.00
|
|
|||||
Description
Rat Monoclonal Antibody
CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.Specifications
| CD205 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 17076 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| 205yekta | |
| Flow Cytometry | |
| Super Bright 600 | |
| Rat | |
| Mouse | |
| Q60767 | |
| LY75 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| LY75 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title