Learn More
Invitrogen™ CD205 Monoclonal Antibody (HD30), PE

Description
This antibody recognizes an extracellular epitope of CD205, an approx. 200 kDa C-type lectin transmembrane protein of the MMR (macrophage mannose receptor) family, expressed at high levels on dendritic cells and thymic epithelial cells, and at low levels on lymphocytes, NK cells and monocytes.
Specifications
Specifications
| Antigen | CD205 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HD30 |
| Conjugate | PE |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | LY75 |
| Gene Accession No. | O60449 |
| Gene Alias | CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4 |
| Gene Symbols | LY75 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.