Learn More
CD205 Mouse anti-Human, Brilliant Violet 421, Clone: MMRI-7, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744056
Description
The MMRI-7 monoclonal antibody specifically recognizes CD205 which is also known as DEC-205, Lymphocyte antigen 75 (Ly-75), or C-type lectin domain family 13 member B (CLEC13B). CD205 is a ñ205 kDa type I transmembrane glycoprotein with C-type lectin external domains. DEC-205 is expressed on myeloid and plasmacytoid dendritic cells, thymic epithelial cells, and at lower levels on monocytes, T cells, B cells, and natural killer (NK) cells. DEC-205 likely functions as an endocytic receptor and is involved in antigen uptake and processing.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD205 | |
| MMRI-7 | |
| Brilliant Violet 421 | |
| DEC-205, Lymphocyte Antigen 75, LY75, Ly-75, CLEC13B, GP200-MR6 | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.