Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD205 Mouse, anti-Human, R718, Clone: MMRI-7, BD OptiBuild™

Catalog No. BDB752288 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752288 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752288 Supplier BD Biosciences Supplier No. 752288
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MMRI-7 monoclonal antibody specifically recognizes CD205 which is also known as DEC-205, Lymphocyte antigen 75 (Ly-75), or C-type lectin domain family 13 member B (CLEC13B). CD205 is a ∼205 kDa type I transmembrane glycoprotein with C-type lectin external domains. DEC-205 is expressed on myeloid and plasmacytoid dendritic cells, thymic epithelial cells, and at lower levels on monocytes, T cells, B cells, and natural killer (NK) cells. DEC-205 likely functions as an endocytic receptor and is involved in antigen uptake and processing.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD205
Applications Flow Cytometry
Classification Monoclonal
Clone MMRI-7
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.