Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD206 (MMR) Monoclonal Antibody (15-2)
CD206 (MMR) Monoclonal Antibody (15-2)
Supplier: Thermo Scientific MA528581
Description
CD206 (MMR) Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), Flow, IP, FN

Specifications
CD206 (MMR) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P22897 | |
Mrc1 | |
Purified human mannose receptor. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
15-2 | |
Unconjugated | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Mouse | |
Protein A | |
RUO | |
4360 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction