Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD206 (MMR) Mouse anti-Human, BV605, Clone: 15-2 (also known as MR15-2), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 751775
Description
CD206 (MMR) Monoclonal antibody specifically detects CD206 (MMR) in Human samples. It is validated for Flow Cytometry.

Specifications
CD206 (MMR) | |
Monoclonal | |
0.2 mg/mL | |
Hu CD206 (MMR) BV605 15-2 50 μg | |
Purified Human MMR | |
RUO | |
CD206 (MMR) | |
Refrigerate (2 to 8°C) | |
Human |
Flow Cytometry | |
15-2 (also known as MR15-2) | |
Brilliant Violet 605 | |
Mouse | |
1 x 50 μg | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction