Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD206 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA583759
Description
Immunogen sequence: YCGELKGDPT MSWNDINCEH LNNWICQIQK GQTPKPEPTP APQDNPPVTE DGWVIYKDYQ YYFSKEKETM DNARAFCKRN FGDLVSIQSE SEKKFLWKYV NRNDAQSA
MA1-35936 recognizes human mannose receptor, also know as CD206, which is a 175kD transmembrane protein belonging to the group of pattern recognition receptors. CD206 is predomitly expressed in tissue macrophages and dentritic cells and can also be found in a subpopulation of endothelial cells and sperm cells. CD206 is thought to play a role in the innate and adaptive immune response.
Specifications
CD206 | |
Polyclonal | |
Unconjugated | |
MRC1 | |
bA541I19.1, CD206, CLEC13D, CLEC13DL, MRC1L1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P22897 | |
MRC1 | |
Recombit protein corresponding to Human MRC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction