Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD206 Rat anti-Mouse, Alexa Fluor 647, Clone: MR5D3, BD Pharmingen™

Catalog No. BDB565250 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565250 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565250 Supplier BD Biosciences Supplier No. 565250
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD206 Monoclonal antibody specifically detects CD206 in Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD206
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone MR5D3
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias MMR; MR; Macrophage mannose receptor 1; Mrc1; Mannose receptor, C type 1
Host Species Rat
Immunogen Recombinant Mouse CD206 Carbohydrate recognition domains 4-7/Fc Fusion Protein
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed.
Form Purified
Isotype IgG2a, CDF IgG2a
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.