Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD207 (Langerin) Monoclonal Antibody (eBioRMUL.2), eBioscience™, Invitrogen™

Catalog No. 14207337
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-207-337 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-207-337 Supplier Invitrogen™ Supplier No. 14207337
Add to Cart
Add to Cart

Rat Monoclonal Antibody

RMUL.2 is a MAb that reacts with the predicted extracellular domain of mouse CD207, also known as Langerin. Mouse CD207 is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). Mouse CD207 is a 48 kDa, C-type lectin and putative transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation reduce expression of CD207 and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines. Western Blotting with RMUL.2 reveals a 48 kDa band in lysate from mouse ear epidermis as well as in lysates from cells transfected with mouse CD207 cDNA. Cross reactivity of this antibody with other proteins has not been determined. Preliminary data from mouse CD207 cDNA transfected cells suggests that RMUL.2 can be used for intracellular staining followed by flow cytometric analysis. We have observed in our transfectants the majority of protein is expressed intracellularly with a small percentage on the cell surface. In addition, RMUL.2 has also been tested by immunohistology on mouse ear epidermal sheets and shows specific staining.

Langerin (CD207) is a type II membrane-associated C-type lectin known to be expressed exclusively by Langerhans cells. Lamgerin recognizes mannose residues via its single carbohydrate recognition domain (CRD). Langerin is localized not only on the cell surface, but also intracellularly in close association with Birbeck granules. Transfection of Langerin cDNA into fibroblasts creates a compact network of membrane structures with typical features of Birbeck granules (BG). Langerin is thus a potent inducer of membrane superimposition and zippering leading to BG formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD207 (Langerin)
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone eBioRMUL.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD207
Gene Accession No. Q8VBX4
Gene Alias CD 207 antigen; CD207; CD207 antigen; CD207 antigen, langerin; CD207 molecule; CD207 molecule, langerin; CLEC4K; C-type lectin domain family 4 member K; C-type lectin domain family 4, member K; Langerhans cell specific c-type lectin; langerin; RGD1565913
Gene Symbols CD207
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246278
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.