Learn More
CD209 (DC-SIGN) Monoclonal Antibody (eB-h209), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$418.00
Specifications
| Antigen | CD209 (DC-SIGN) |
|---|---|
| Clone | eB-h209 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3107
![]()
|
Invitrogen™
17209942 |
100 Tests |
Each of 1 for $418.00
|
|
|||||
Description
Description: The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44 kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN. Applications Reported: The eB-h209 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eB-h209 antibody has been pre-titrated and tested by flow cytometric analysis of cultured human dendritic cells and peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD209 (DC-SIGN) Monoclonal antibody specifically detects CD209 (DC-SIGN) in Human samples. It is validated for Flow Cytometry.Specifications
| CD209 (DC-SIGN) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q9NNX6 | |
| CD209 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD209 |
| eB-h209 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Human | |
| 30835 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.