Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD209a Rat anti-Mouse, BV605, Clone: 5H10, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747825
Description
CD209a Monoclonal antibody specifically detects CD209a in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD209a | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
5H10 | |
Brilliant Violet 605 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction