Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD209b Rat anti-Mouse, BV421, Clone: ER-TR9 (also known as ER-TR9), BD Horizon™
Rat Monoclonal Antibody
$104.50 - $253.50
Specifications
Antigen | CD209b |
---|---|
Clone | ER-TR9 (also known as ER-TR9) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD209b Monoclonal antibody specifically detects CD209b in Mouse samples. It is validated for Flow cytometrySpecifications
CD209b | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
69165 | |
IgM κ | |
Affinity Chromatography |
ER-TR9 (also known as ER-TR9) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Rat | |
Mouse | |
Cd209b; DC-SIGN-related protein 1; DC-SIGNR1; SIGNR1; mSIGNR1 | |
C3H Mouse Thymic Stromal Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title