Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD209b (SIGN-R1) Hamster anti-Mouse, BV480, Clone: 22D1, BD OptiBuild™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 756166
Description
CD209b (SIGN-R1) Monoclonal antibody specifically detects CD209b (SIGN-R1) in Mouse samples. It is validated for Flow cytometry
Specifications
CD209b (SIGN-R1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
69165 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG κ |
Flow Cytometry | |
22D1 | |
Brilliant Violet 480 | |
Cd209b; DC-SIGNR1; mSIGNR1 | |
Recombinant Mouse SIGN-R1 ECD–Mouse IgG-Fc Protein | |
50 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction