Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Antibody (21A/5), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250178AF750
Description
CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
CD21 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CR2 | |
CD21 | |
0.1 mL | |
1380 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry | |
21A/5 | |
Flow Cytometry, Immunohistochemistry | |
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction