Learn More
CD21 Antibody (BB6-11C9.6), Alexa Fluor™ 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128245AF405
Catalog No. NBP128245W
Only null left
Add to Cart
Description
Description
CD21 Monoclonal specifically detects CD21 in Mouse, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD21
Monoclonal
Alexa Fluor 405
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Mouse
Protein A or G purified
RUO
Primary
Porcine CD21, Mr 160 kDa
Store at 4C in the dark.
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
BB6-11C9.6
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CR2
Porcine CD21, Mr 160 kDa
0.25 mL
Adaptive Immunity, Dendritic Cell Markers, Immunology
1380
Mouse, Pig
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only