Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Antibody (Bu33), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063574V
Description
CD21 Monoclonal specifically detects CD21 in Human, Equine samples. It is validated for Flow Cytometry.Specifications
CD21 | |
Monoclonal | |
DyLight 405 | |
P20023 | |
CR2 | |
Made to Human CD21 | |
0.1 mL | |
Adaptive Immunity, Dendritic Cell Markers, Immunology | |
1380 | |
Human, Equine | |
Purified |
Flow Cytometry | |
Bu33 | |
Flow Cytometry | |
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-63574 recognizes human CD21, a 145kD cell surface glycoprotein expressed by mature B lymphocytes, follicular dendritic cells, subsets of normal thymocytes and subsets of T cells. The CD21 molecule acts as a receptor for CD23, for complement components C3d, C3dg and iC3b and for Epstein Barr Virus. CD21 is part of the signalling complex that is linked to CD19, CD81 and phosphatidylinositol-3-kinase. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction