Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD21/CD35 Monoclonal Antibody (eBio8D9 (8D9)), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010264
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-102-64 100 μg
50-102-63 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-102-64 Supplier Invitrogen™ Supplier No. 12021182
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody eBio8D9 reacts with an eptiope shared by mouse CD21 (CR2) and CD35 (CR1). CD21 and CD35 are alternatively spliced transcripts from the Cr2 gene, which produce cell-surface proteins of 145 and 190 kDa, respectively. CD21 and CD35 are expressed by mature B cells, but not on thymocytes, peripheral T cells, erythrocytes or platelets. Furthermore, there is some evidence which demonstrates their expression on macrophages. CD21 is a receptor for the complement component C3d and Epstein-Barr virus (EBV). In association with CD19 and CD81, CD21 also participates in B-cell activation through the B cell receptor. Cr2-deficient mice display impaired inflammatory and humoral immune responses in vivo. The anti-mouse CD21/35 monoclonal antibody clones eBio4E3 and eBio8D9 do not cross-block each other, suggesting that they bind to different epitopes. Applications Reported: This 4E3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio8D9 (8D9) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

CD21 (complement receptor 2, CR2) binds C3 complement fragments, especially its breakdown fragments, which remain covalently attached to complement activating surfaces or antigen. CD21 has important roles in uptake and retention of immunocomplexes, survival of memory B cells and in development and maintenance of the humoral response to T-dependent antigens. CD21 also serves as a key receptor for Epstein-Barr virus binding and is involved in targeting prions to folicular dendritic cells and expediting neuroinvasion following peripheral exposure to prions. A soluble form of the CD21 (sCD21) is shed from the lymphocyte surface and retains its ability to bind respective ligands. CD35 encodes a membrane glycoprotein found on peripheral blood cells, glomerular podocytes, and follicular dendritic cells. The protein is a receptor for complement components C3b and C4b and regulates the activity of the complement cascade. Variation in this protein is the basis of the Knops blood group system. The two most common alleles, F and S, differ by 8 exons and are thought to be the result of an unequal crossover event. A secreted form of the protein present in plasma has been described, but its full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD21/CD35
Applications Flow Cytometry
Classification Monoclonal
Clone eBio8D9 (8D9)
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cr2
Gene Accession No. P19070
Gene Alias C3b/C4b receptor; C3-binding protein; C3DR; CD21; CD35; CD35 antigen; Complement C3d receptor; complement component (3d/Epstein Barr virus) receptor 2; complement component 3d receptor 2; complement receptor 2; complement receptor type 1; complement receptor type 2; CR; Cr1; Cr-1; Cr2; Cr-2; CVID7; EBV receptor; Epstein-Barr virus receptor; expressed in B lymphocytes; SLEB9
Gene Symbols Cr2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12902
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.