Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Monoclonal Antibody (HB5), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD21 |
---|---|
Clone | HB5 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H055T03B06A
![]() |
Invitrogen™
H055T03B06A |
100 Tests |
Each for $430.00
|
|
|||||
H055T02B06A
![]() |
Invitrogen™
H055T02B06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Flow CytometrySpecifications
CD21 | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P20023 | |
1380 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HB5 | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
IgG2a κ | |
Affinity chromatography | |
CR2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title