Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD210a Rat anti-Human, Brilliant Violet 421, Clone: 3F9, BD Optibuild™

Catalog No. BDB742942 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742942 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742942 Supplier BD Biosciences Supplier No. 742942
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 3F9 monoclonal antibody specifically binds to CD210a, which is also known as, Interleukin-10 Receptor subunit alpha (IL-10R subunit alpha/IL-10R /IL10RA), or Interleukin-10 receptor subunit 1 (IL-10R1). CD210a is a 90-110 kDa type I transmembrane glycoprotein that belongs to the type II cytokine receptor family. CD210a combines with IL-10 Receptor subunit beta (IL-10R /IL10RB/CDw210b) to form the IL-10 Receptor complex ( ) that can bind IL-10 and transduce signals intracellularly through the JAK/STAT pathway. IL-10 can suppress antigen presentation and the expression of proinflammatory type-1 immune responses while promoting type-2 immune responses. CD210a is expressed on T cells, B cells, NK cells, monocyte, macrophages and dendritic cells. Clone 3F9 is specific for human CD210a and its binding to the receptor can be blocked by recombinant human IL-10 (rhIL-10) protein.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD210a
Classification Monoclonal
Clone 3F9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD210, CDW210A, IL10RA, IL-10RA, IL-10R-A, IL10R, HIL-10R, IL-10R1, I10R1
Host Species Rat
Immunogen Human IL-10R alpha Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.