Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD215 (IL-15Rα) Mouse anti-Human, APC, Clone: JM7A4 (also known as 7A4), BD Pharmingen™
Mouse Monoclonal Antibody
$170.00 - $348.00
Specifications
Antigen | CD215 (IL-15Rα) |
---|---|
Clone | JM7A4 (also known as 7A4) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
CD215 (IL-15Rα) Monoclonal antibody specifically detects CD215 (IL-15Rα) in Human samples. It is validated for Flow CytometrySpecifications
CD215 (IL-15Rα) | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
IL15RA; IL-15R-alpha; IL-15RA; IL-15Rα; CD215 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
JM7A4 (also known as 7A4) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Recombinant Human IL-15Rα Extracellular Domain | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title