Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD215 (IL-15Rα) Mouse anti-Human, BV711, Clone: JM7A4 (also known as 7A4), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747700
Description
CD215 (IL-15Rα) Monoclonal antibody specifically detects CD215 (IL-15Rα) in Human samples. It is validated for Flow Cytometry.
Specifications
CD215 (IL-15Rα) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
JM7A4 (also known as 7A4) | |
Brilliant Violet 711 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction