Learn More
Invitrogen™ CD215 (IL-15Ra) Monoclonal Antibody (eBioJM7A4), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 14715937
Description
The eBioJM7A4 MAb reacts with the IL-15 receptor alpha chain (IL-15Ralpha). IL-15 and IL-2 are cytokines with overlapping, but distinct, biologic effects. Their receptors share 2 subunits, the IL-2Rbeta and gamma chains, which are essential for signal transduction. The IL-15Ralpha receptor is structurally related to IL-2Ralpha. However, the IL-15Ralpha alone binds IL-15 with a 1,000-fold higher affinity than that seen with IL-2Ralpha and IL-2. In the human, 3 differentially spliced human IL-15Ralpha variants that are all capable of high affinity binding of IL-15 have been isolated. The cytoplasmic domain of IL-15Ralpha, like that of IL-2Ralpha, is dispensable for mitogenic signaling, suggesting that the primary role of the alpha chains is to confer high affinity binding. At high concentrations, IL-15, like IL-2, is able to signal through a complex of IL-2Rbeta and IL-2Rgamma in the absence of the alpha subunit.
Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
Specifications
| CD215 (IL-15Ra) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q13261 | |
| IL15RA | |
| Affinity chromatography | |
| RUO | |
| 3601 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| eBioJM7A4 | |
| Unconjugated | |
| IL15RA | |
| AA690181; CD215; IL 15 receptor; IL 15 receptor subunit α; IL 15R; IL 15R alpha; IL 15R α; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit α; IL15R; IL15R alpha; IL15R α; Il15ra; IL-15RA; IL-15R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit α; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15R; sIL-15 receptor subunit alpha; sIL15R; sIL-15RA; sIL-15R-alpha; soluble IL 15R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha | |
| Mouse | |
| 2 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.