Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD217 (IL-17RA) Mouse anti-Human, BUV615, Clone: 133617, BD OptiBuild™

Catalog No. BDB752336 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752336 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752336 Supplier BD Biosciences Supplier No. 752336
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD217 (IL-17RA) Monoclonal antibody specifically detects CD217 (IL-17RA) in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD217 (IL-17RA)
Applications Flow Cytometry
Classification Monoclonal
Clone 133617
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu CD217 (IL-17RA) BUV615 133617 50 μg
Host Species Mouse
Immunogen Human CD217 (IL-17RA) Recombinant Protein
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD217 (IL-17RA)
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG1 κ
Reactivity Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.