Learn More
Description
Host Species: Mouse
Clone: FPC1
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant protein encoding the external domain of human CD22
Molecular Weight: 130-140kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Specifications
Specifications
| Antigen | CD22 Ab-4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | FPC1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant Protein encoding the external domain of human CD22 |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | B-Cell |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.