Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD221 Mouse anti-Human, PE, Clone: 1H7, BD

Catalog No. BDB555999 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555999 100 Tests
BDB560934 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555999 Supplier BD Biosciences Supplier No. 555999
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1H7 monoclonal antibody specifically binds to the α subunit of the human insulin-like growth factor-I (IGF-I) receptor that is also known as CD221. CD221 is a receptor tyrosine kinase that has high affinity for IGF-1 and low affinity for insulin and IGF-2. CD221 is synthesized as a 180 kDa precursor molecule that is processed into two subunits, a 135 kDa α subunit which is disulfide-linked to a 90 kDa β subunit. The mature IGF-IR complex is comprised of two extracellular α subunits and two transmembrane β subunits that each possesses a tyrosine kinase domain. When bound by ligand, the IGF-1 receptor autophosphorylates multiple tyrosine sites in its β chains and phosphorylates other signaling molecules that regulate cell growth, development, and neoplastic transformation. CD221 is structurally and functionally similar to the insulin receptor (aka, CD220) and is expressed on a variety of human hematopoietic and non-hematopoietic cells. The 1H7 antibody does not crossreact with the human insulin receptor and can block IGF-I- or IGF-II-stimulated cell growth.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD221
Applications Flow Cytometry
Classification Monoclonal
Clone 1H7
Conjugate PE
Description IGF1R; IGF-I receptor; IGFIR; IGFR; Insulin-like growth factor 1 receptor
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Human Placental IGF-I Receptor
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.