Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD222 Mouse, anti-Human, R718, Clone: MEM-238, BD OptiBuild™

Catalog No. BDB751976 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751976 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751976 Supplier BD Biosciences Supplier No. 751976
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MEM-238 monoclonal antibody specifically binds to CD222, which is also known as the Insulin-like growth factor 2 Receptor (IGF2R, IGF-II Receptor), Cation-independent mannose 6-phosphate receptor (CIMPR), or Mannose-6 phosphate receptor (M6PR). CD222 is ubiquitously expressed by a variety of cell types as a cell surface type I transmembrane glycoprotein. However, in the course of receptor trafficking between the cell surface and intracellular compartments, the majority of CD222 is found within cells. Cell surface CD222 functions as a multifunctional receptor that binds to a large number of extracellular ligands including acid hydrolases, insulin-like growth factors, latent TGF-β, leukemia inhibitory factor (LIF), proliferin, prorenin, plasminogen, and Herpes simplex virus. It regulates extracellular Insulin-like growth factor II (IGF-II/IGF-2) levels by binding and internalizing the growth factor for lysosomal degradation. This effectively removes IGF-II from the circulation and tissues and thus prevents it from signaling through the growth-stimulatory Insulin-like growth factor I Receptor (IGF-1R, CD221) pathway. However, studies have reported that IGF-II may activate some cellular functions through CD222 as well. A soluble form of the CD222 extracellular region can also be detected in human serum and may play a role in regulating IGF-II activity. CD222 serves as a surface receptor for latent TGFβ and can complex with plasminogen and CD87, a urokinase-type plasminogen activator receptor, to activate latent TGF-β. CD222 also binds to a variety of Mannose 6-phosphate (M6P)-containing proteins, including extracellular and newly-synthesized lysosomal enzymes, and transports them to lysosomes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD222
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-238
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.