Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD223 (LAG-3) Monoclonal Antibody (3DS223H), NovaFluor™ Yellow 590, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD223 (LAG-3) |
---|---|
Clone | 3DS223H |
Concentration | 0.4 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H048T03Y02A
![]() |
Invitrogen™
H048T03Y02A |
100 Tests |
Each for $430.00
|
|
|||||
H048T02Y02A
![]() |
Invitrogen™
H048T02Y02A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD223 (LAG-3) Monoclonal antibody specifically detects CD223 (LAG-3) in Human samples. It is validated for Flow CytometrySpecifications
CD223 (LAG-3) | |
0.4 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P18627 | |
3902 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
3DS223H | |
Flow Cytometry | |
NovaFluor Yellow 590 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
IgG1 κ | |
Affinity chromatography | |
LAG3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title