Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD223 (LAG-3) Recombinant Rabbit Monoclonal Antibody (BLR028F)

Catalog No. PIMA544252
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544252 10 μL
PIMA544251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544252 Supplier Invitrogen™ Supplier No. MA544252
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

LAG-3 is a 70-kDa surface glycoprotein in the immunoglobulin superfamily, with structural homology to CD4. It binds to MHC class II molecules with higher affinity than CD4 and is involved in the negative regulation of T cell activation and homeostatic proliferation. LAG-3 is expressed on the surface of activated T cells, including regulatory T cells, and NK cells. Notably, CD8+ T cells express LAG-3 at significantly higher levels than CD4+ T cells. Additionally, the coexpression of LAG-3 and CD49b has been proposed as a marker for identifying Type 1 regulatory T cells (Tr1 cells) in both humans and mice. This highlights LAG-3s role in immune regulation and its potential as a target for therapeutic interventions in immune-related conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD223 (LAG-3)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR028F
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.09% sodium azide; pH 8.2
Gene LAG3
Gene Accession No. P18627
Gene Alias Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3
Gene Symbols LAG3
Host Species Rabbit
Immunogen Recombinant protein representing the extracellular domain of human Lymphocyte activation gene 3 (LAG3, residues 23-450).
Purification Method Purified
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.