Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD223 Mouse anti-Human, Alexa Fluor 647, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565716
Description
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.
CD223 Monoclonal antibody specifically detects CD223 in Human samples. It is validated for Flow cytometry.
Specifications
CD223 | |
Monoclonal | |
Alexa Fluor 647 | |
LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
Human LAG-3 Recombinant Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
IgG1 κ |
Flow Cytometry | |
T47-530 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction