Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD223 Recombinant Llama Monoclonal Antibody (2A11)
Llama Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549322
Description
CD223 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA
LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Specifications
CD223 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P18627 | |
LAG3 | |
A recombinant protein corresponding to amino acids 23-434 of human LAG3. | |
100 μg | |
Primary | |
Human | |
Antibody | |
VHH |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
2A11 | |
Unconjugated | |
LAG3 | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
Llama | |
Affinity chromatography | |
RUO | |
3902 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction