Learn More
Invitrogen™ CD223 Recombinant Llama Monoclonal Antibody (2B11)

Description
CD223 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA
Specifications
Specifications
| Antigen | CD223 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 2B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | LAG3 |
| Gene Accession No. | P18627 |
| Gene Alias | Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.