Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD226 (DNAM-1) Rat anti-nan, BV421, Clone: TX42.1 (also known as TX42), BD Horizon™
Rat Monoclonal Antibody
$188.80
Specifications
Antigen | CD226 (DNAM-1) |
---|---|
Clone | TX42.1 (also known as TX42) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD226 (DNAM-1) Monoclonal antibody specifically detects CD226 (DNAM-1) in nan samples. It is validated for Flow cytometrySpecifications
CD226 (DNAM-1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
CD226 antigen; Cd226; DNAM-1; DNAM1; Pta1; TLiSA1 | |
Mouse DNAM-1 transfected cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
TX42.1 (also known as TX42) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Rat | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
225825 | |
IgG2a κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title